Hydroxypropyl methyl cellulose, M.N. 86,000, viscosity 4,000 cP (2% solution)
Hydroxypropyl methyl cellulose, M.N. 86,000, viscosity 4,000 cP (2% solution)
Hydroxypropyl methyl cellulose, M.N. 86,000, viscosity 4,000 cP (2% solution)
Hydroxypropyl methyl cellulose, M.N. 86,000, viscosity 4,000 cP (2% solution)
Thermo Scientific Chemicals

Hydroxypropyl methyl cellulose, M.N. 86,000, viscosity 4,000 cP (2% solution)

CAS: 9004-65-3 | C56H108O30 | 1261.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 244021000
Price (USD)
99.65
Special offer
Online exclusive
Ends: 26-Sep-2025
123.00
Save 23.35 (19%)
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 26-Sep-2025
123.00
Save 23.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS9004-65-3
IUPAC Name1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular FormulaC56H108O30
InChI KeyPUSNGFYSTWMJSK-UHFFFAOYNA-N
SMILESCOCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<10 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link