Pierce™ Glycoprotein Staining Kit
Pierce™ Glycoprotein Staining Kit
Thermo Scientific™

Pierce™ Glycoprotein Staining Kit

The Pierce Glycoprotein Staining Kit is a convenient, fast, and sensitive colorimetric kit for staining glycosylated proteins in polyacrylamide gelsRead more
Have Questions?
Catalog NumberQuantity
24562750 mL
Catalog number 24562
Price (EUR)
421,00
Each
-
Add to cart
Quantity:
750 mL
Request bulk or custom format
Price (EUR)
421,00
Each
Add to cart
Ask our AI about this Product
The Pierce Glycoprotein Staining Kit is a convenient, fast, and sensitive colorimetric kit for staining glycosylated proteins in polyacrylamide gels or nitrocellulose membranes using the periodic acid-Schiff (PAS) method.

Features:
Specific for glycoproteins—PAS method specifically detects glycosylated proteins with sialic acid and other oxidizable carbohydrate groups
Protocol—the traditional three-reagent method has been streamlined to achieve optimal staining in less than two hours
Colorimetric—glycoproteins are detected as magenta bands against a faint pink or colorless gel background
Qualitative staining—detection limit depends on the extent of glycosylation and protein size and abundance; detects 0.625 ng avidin and 0.16 μg horseradish peroxidase per band
Controls included—complete kit includes positive and negative control proteins (HRP and STI, respectively).

Compare all glycoprotein stains ›

The kit provides three essential reagents and a complete protocol to specifically stain glycosylated proteins (glycoproteins) that have been separated by polyacrylamide gel electrophoresis (PAGE). A gel or membrane containing separated proteins is treated with a periodate solution, which oxidizes cis-diol sugar groups in glycoproteins. The resulting aldehyde groups are detected through the formation of Schiff-base bonds with a reagent that produces magenta bands. Traditionally called the periodic acid-Schiff (PAS) reagent method, this staining technique is made easier by the convenient packaging of essential reagents in one kit with easy-to-follow instructions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection LocationIn-Blot Detection, In-Gel Detection
Detection MethodColorimetric
Product LinePierce™
Product TypeGlycoprotein Staining Kit
Quantity750 mL
Target MoleculeProteins (Glycoproteins)
Label or DyeFuchsin Sulfite
Unit SizeEach
Contents & Storage
Sufficient For: 10 mini gels (SDS-PAGE)
• Oxidation Reagent, 2.5 g powder to make 250 mL
• Reduction Reagent, 1.25 g powder to make 250 mL
• Positive Control (Horseradish Peroxidase), 1 mg
• Negative Control (Soybean Trypsin Inhibitor), 1 mg
• Glycoprotein Stain, 250 mL

Store all components at 4°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link