Thermo Scientific Chemicals

Triisopropanolamine, 98%, Thermo Scientific Chemicals

Catalog number: 245761000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Triisopropanolamine, 98%, Thermo Scientific Chemicals

Catalog number: 245761000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 245761000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-20-3
IUPAC Name
1-[bis(2-hydroxypropyl)amino]propan-2-ol
Molecular Formula
C9H21NO3
InChI Key
SLINHMUFWFWBMU-UHFFFAOYNA-N
SMILES
CC(O)CN(CC(C)O)CC(C)O
Appearance (Color)
White to light yellow
Appearance (Form)
Low melting crystalline solid
Infrared spectrum
Conforms
Titration with HCl
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text