Thermo Scientific™

Pierce™ Zinc Reversible Stain Kit

Catalog number: 24582
Have Questions?
Thermo Scientific™

Pierce™ Zinc Reversible Stain Kit

Catalog number: 24582
Have Questions?
Catalog Number
24582
Unit Size
1.5 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
245821.5 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Zinc Reversible Stain Kit provides rapid and highly sensitive detection of protein in polyacrylamide gels immediately after electrophoresis as well as erasability for subsequent transfer or band extraction.

Features of Zinc Reversible Stain Kit:

Sensitive—detect 0.25ng of protein per band in a mini gel
Fast—the entire protocol is complete in 15 minutes
Easy—ready-to-use stain and developer solutions are supplied, and no fixing or washing steps are required
Gentle—no chemical modification of the proteins occurs; protein bands are visible because the background is stained, not the bands themselves
Reversible—completely erase the stain from the gel to easily recover proteins or proceed with other methods

Includes:
• Zinc stain, developer, and eraser solutions (500 mL each)

Applications:
• Staining preceding protein recovery for antibody generation or immunological detection
• Staining before excising gel pieces for protein digestion and analysis by mass spectrometry
• Staining before electrophoretic transfer to membrane for Western blotting
• Quick purity checks before proceeding with purification or transfer for Western blotting

Pierce Zinc Reversible Stain Kit is an ideal option for in-gel protein detection preceding many downstream applications, because the zinc-stain is completely reversible. The white background is easily removed and the gel is restored to its original condition with the kit-supplied eraser solution or a Tris-glycine buffer. Afterwards, the proteins are free of any dyes or other contaminants and can be restained by another method, transferred to membrane for Western blotting or eluted from an excised gel piece.

Zinc-based protein gel stain provides rapid detection of proteins after their separation by SDS-PAGE. The method is as sensitive as most silver staining techniques in that it detects protein at less than 1ng per band, yet requires less than 15 minutes to complete. The simple, two-step staining protocol results in a stained gel with an opaque milky-white background and clear protein bands. The clear bands of protein are visible when the gel is placed against a dark background.

Related Products
Pierce™ Zinc Reversible Stain Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Colorimetric
Label or Dye
Proprietary Mix
Product Type
Reversible Protein Gel Stain Kit
Product Line
Pierce™
Target Molecule
Protein

Contents & Storage

Sufficient For: 20 mini gels (SDS-PAGE)
• Zinc Stain, 500 mL
• Zinc Developer, 500 mL
• Zinc Eraser, 500 mL

Store kit at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text