Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | GelCode™ Blue Stain Reagent, 3.5 L |
Detection Location | In-Blot Detection, In-Gel Detection |
Detection Method | Colorimetric |
Label or Dye | Coomassie |
Product Type | Protein Gel Stain Reagent |
Product Line | GelCode™ |
Quantity | 3.5 L |
Target Molecule | Protein |
Unit Size | 3.5 L |
Description | GelCode™ Blue Stain Reagent, 500 mL |
Detection Location | In-Blot Detection, In-Gel Detection |
Detection Method | Colorimetric |
Label or Dye | Coomassie |
Product Type | Protein Gel Stain Reagent |
Shelf Life | 6 Months |
Product Line | GelCode™ |
Quantity | 500 mL |
Target Molecule | Protein |
Unit Size | 500 mL |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
24592 | 3.5 L | GelCode™ Blue Stain Reagent, 3.5 L | 387.65 Online price 421.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
24590 | 500 mL | GelCode™ Blue Stain Reagent, 500 mL | 112.65 Online price 122.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
1. Mateer, S.C.,et al.(2002).J. Biol. Chem.227,12324-12333.
2. Aulak, K.S.,et al.(2001).Proc. Natl. Acad. Sci. USA98,12056-12061.
3. Lim, J.,et al.(2002).J. Biol. Chem.277,20774-20782.
4. Hilton, J.M.,et al.(2001).J. Biol. Chem.276,16341-16347.
5. Tani, M.,et al.(2002).J. Biol. Chem.275,3462-3468.