Thermo Scientific™

Imperial™ Protein Stain

Catalog number: 24615
Have Questions?
Thermo Scientific™

Imperial™ Protein Stain

Catalog number: 24615
Have Questions?
Catalog Number
24615
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
3 x 1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
246151 L
-
246173 x 1 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Imperial Protein Stain is a ready-to-use colorimetric stain formulated with coomassie dye R-250 that delivers consistent nanogram-level detection of proteins in polyacrylamide electrophoresis gels or nitrocellulose membranes.

Features:
Sensitive—detects less than 3 ng protein per band with the enhanced protocol (3 hours)
Time to stain—ready-to-use reagent detects less than 6 ng protein per band in just 20 minutes
High contrast—intense purple bands are easier to photograph or scan than typical coomassie blue stains
Compatibility—compatible with downstream mass spectrometry analysis and protein sequencing
Additional reagents—water washes only; no acid-fixative or methanol destaining solutions required
Stable—1-year room-temperature stability

Compare all Coomassie stains ›

The stain is a unique formulation of coomassie brilliant blue R-250 that delivers substantial improvements in protein-staining performance compared to homemade or other commercial stains. Staining results in intensely colored protein bands that are easy to photograph and document with gel imagers. This reagent is one of the most sensitive colorimetric stains available, easily detecting 3 to 6 nanograms of protein per band. The easy-to-follow protocol is flexible to meet demanding time and sensitivity requirements.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Blot Detection, In-Gel Detection
Detection Method
Colorimetric
Label or Dye
Coomassie
Product Type
Protein Stain
Shelf Life
1 Year
Product Line
Imperial™
Quantity
1 L
Target Molecule
Protein

Contents & Storage

Upon receipt store product at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text