Thermo Scientific Chemicals

Sodium bromide, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 246901000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium bromide, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 246901000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 246901000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-15-6
IUPAC Name
sodium bromide
Molecular Formula
BrNa
InChI Key
JHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES
[Na+].[Br-]
pH
5 to 8.8 (5 % at 20°C)
Chloride (Cl)
=<2000 ppm
Bromate (BrO3)
=<10 ppm
Iron (Fe)
=<10 ppm
Barium (Ba)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text