Thermo Scientific Chemicals

2-Hydroxynicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: 247221000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydroxynicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: 247221000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 247221000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
609-71-2
IUPAC Name
2-oxo-1,2-dihydropyridine-3-carboxylic acid
Molecular Formula
C6H5NO3
InChI Key
UEYQJQVBUVAELZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CNC1=O
Appearance (Color)
White to cream to light yellow
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Melting point
256°C to 262°C
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text