Thermo Scientific Chemicals

(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific Chemicals

Catalog number: 247735000
500 mg, Each
Thermo Scientific Chemicals

(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific Chemicals

Catalog number: 247735000
500 mg, Each
Quantity
Catalog number: 247735000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19132-06-0
IUPAC Name
(2S,3S)-butane-2,3-diol
Molecular Formula
C4H10O2
InChI Key
OWBTYPJTUOEWEK-IMJSIDKUSA-N
SMILES
C[C@H](O)[C@H](C)O
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %
Infrared spectrum
Conforms
Specific optical rotation
+12° to +14° (20°C, 589 nm) (neat)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text