Thermo Scientific Chemicals

Ethyl potassium malonate, 98%, Thermo Scientific Chemicals

Catalog number: 248081000
100 g, Each
Thermo Scientific Chemicals

Ethyl potassium malonate, 98%, Thermo Scientific Chemicals

Catalog number: 248081000
100 g, Each
Quantity
Catalog number: 248081000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6148-64-7
IUPAC Name
3-ethoxy-3-oxopropanoic acid potassium
Molecular Formula
C5H8KO4
InChI Key
HRBCMCJOBRIIQG-UHFFFAOYSA-N
SMILES
[K].CCOC(=O)CC(O)=O
Appearance (Color)
White to cream
Infrared spectrum
Conforms
Titration with HClO4
97.5 to 102.5 %
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text