Thermo Scientific Chemicals

(±)-trans-1,2-Diaminocyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: 248670500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

(±)-trans-1,2-Diaminocyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: 248670500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 248670500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1121-22-8
IUPAC Name
cyclohexane-1,2-diamine
Molecular Formula
C6H14N2
InChI Key
SSJXIUAHEKJCMH-UHFFFAOYNA-N
SMILES
NC1CCCCC1N
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Liquid
Appearance
May develop some turbidity or precipitate
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text