Thermo Scientific Chemicals

Divinyltetramethyldisiloxane, 97%, Thermo Scientific Chemicals

Catalog number: 249210500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Divinyltetramethyldisiloxane, 97%, Thermo Scientific Chemicals

Catalog number: 249210500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 249210500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2627-95-4
IUPAC Name
ethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane
Molecular Formula
C8H18OSi2
InChI Key
BITPLIXHRASDQB-UHFFFAOYSA-N
SMILES
C[Si](C)(O[Si](C)(C)C=C)C=C
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4100 to 1.4130 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text