Thermo Scientific Chemicals

1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals

Catalog number: 249460500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals

Catalog number: 249460500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 249460500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
615-41-8
IUPAC Name
1-chloro-2-iodobenzene
Molecular Formula
C6H4ClI
InChI Key
MPEOPBCQHNWNFB-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1I
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow to orange
GC
>=98.5 %
Refractive index
1.6335 to 1.6365 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text