Thermo Scientific Chemicals

3-Iodophenol, 99%, Thermo Scientific Chemicals

Catalog number: 249700250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Iodophenol, 99%, Thermo Scientific Chemicals

Catalog number: 249700250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 249700250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-02-8
IUPAC Name
3-iodophenol
Molecular Formula
C6H5IO
InChI Key
FXTKWBZFNQHAAO-UHFFFAOYSA-N
SMILES
OC1=CC=CC(I)=C1
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
or fused solid
Melting point
37°C to 43°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text