Methyl 2-bromobenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 2-bromobenzoate, 98%, Thermo Scientific Chemicals
 
Have Questions?
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number 249810500
Price (USD)/ Each
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS610-94-6
IUPAC Namemethyl 2-bromobenzoate
Molecular FormulaC8H7BrO2
InChI KeySWGQITQOBPXVRC-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.5575 to 1.5595 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only