Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Actual product may vary
Thermo Scientific™
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Recover samples completely with these conical well bottom plates. The bottom shape allows for easy pipette access.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityLidNo. per Case
249940SterileWithout Lid80
249570Non-sterileWithout Lid180
249662SterileWithout Lid50
249935SterileWith Lid80
249952SterileWith Lid160
5 Options
Catalog number 249940
Price (USD)/ Case of 80
394.65
Online exclusive
414.00 
Save 19.35 (5%)
-
Add to cart
Sterility:
Sterile
Lid:
Without Lid
No. per Case:
80
Price (USD)/ Case of 80
394.65
Online exclusive
414.00 
Save 19.35 (5%)
Add to cart
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Catalog number249940
Price (USD)/ Case of 80
394.65
Online exclusive
414.00 
Save 19.35 (5%)
-
Add to cart
Recover samples completely with Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates. The bottom shape allows for easy pipette access. The plates are optimized for robotics, automated readers and liquid handling systems.
Features:
  • Conical bottom (V) microplates are ideal for sample recovery.
    • Applications include antibiotic screens, serological tests, storing and screening compounds, DNA libraries, and titration of solutions.
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWithout Lid
Array8 x 12
Packaging80 Cases
ColorClear
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
SterilitySterile
Surface TreatmentNon-treated
Well ShapeConical Bottom (V-well)
No. per Case80
No. per Pack10
Quantity80/Cs.
Unit SizeCase of 80