Search Thermo Fisher Scientific
Catalog Number | Color | Sterility | Well Shape | Fit for Purpose |
---|---|---|---|---|
249944 | Natural | Non-sterile | Round, Conical Bottom (V-well) | Certified Quality |
249943 | Red | Non-sterile | Round, Conical Bottom (V-well) | Certified Quality |
249945 | Black | Non-sterile | Round, Conical Bottom (V-well) | Certified Quality |
249946 | Natural | Sterile | Round, Conical Bottom (V-well) | Sterile |
249947 | Yellow | Non-sterile | Round, Conical Bottom (V-well) | Certified Quality |
249949 | White | Non-sterile | Round, Conical Bottom (V-well) | Certified Quality |
249950 | Blue | Non-sterile | Round, Conical Bottom (V-well) | Certified Quality |
267245 | Natural | Non-sterile | Round | Certified Quality |
267334 | Natural | Sterile | Round | Certified Quality |
267342 | Black | Non-sterile | Round | Certified Quality |
267350 | White | Non-sterile | Round | Certified Quality |
267385 | Blue | Non-sterile | Round | Certified Quality |
267407 | Yellow | Non-sterile | Round | Certified Quality |
442587 | Natural | Non-sterile | Round, Conical Bottom (V-well) | Certified Quality |
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
Gain confidence that the products you buy are ideally suited to your application.
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Nalgene and Nunc products.
Start your request
Share catalog number, name or link