Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Thermo Scientific™
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorWell ShapeSterilityFit for Purpose
249947YellowRound, Conical Bottom (V-well)Non-sterileCertified Quality
249943RedRound, Conical Bottom (V-well)Non-sterileCertified Quality
249944NaturalRound, Conical Bottom (V-well)Non-sterileCertified Quality
249945BlackRound, Conical Bottom (V-well)Non-sterileCertified Quality
249946NaturalRound, Conical Bottom (V-well)SterileSterile
249949WhiteRound, Conical Bottom (V-well)Non-sterileCertified Quality
249950BlueRound, Conical Bottom (V-well)Non-sterileCertified Quality
267245NaturalRoundNon-sterileCertified Quality
267334NaturalRoundSterileCertified Quality
267342BlackRoundNon-sterileCertified Quality
267350WhiteRoundNon-sterileCertified Quality
267385BlueRoundNon-sterileCertified Quality
267407YellowRoundNon-sterileCertified Quality
442587NaturalRound, Conical Bottom (V-well)Non-sterileCertified Quality
Catalog number 249947
Price (USD)/ Case of 120
565.25
Online exclusive
665.00 
Save 99.75 (15%)
-
Add to cart
Color:
Yellow
Well Shape:
Round, Conical Bottom (V-well)
Sterility:
Non-sterile
Fit for Purpose:
Certified Quality
Price (USD)/ Case of 120
565.25
Online exclusive
665.00 
Save 99.75 (15%)
Add to cart
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Catalog number249947
Price (USD)/ Case of 120
565.25
Online exclusive
665.00 
Save 99.75 (15%)
-
Add to cart
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
  • Shared wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
Format96-well
LidWithout Lid
Description96-well Polypropylene Sample Processing & Storage Plates
Array8 x 12
AutoclavableAutoclavable
Packaging20 pk., 120 cs.
Binding TypeLow binding PP
Product LineNunc
ColorYellow
Product Type96-well plate
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
MaterialPolypropylene
Volume (Metric) Well450 μL
No. of Wells96
Volume (Metric) Working400 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeRound, Conical Bottom (V-well)
Dnase FreeYes
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
No. per Case120
No. per Pack20
Quantity120/Cs.
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
RNase FreeYes
Fit for PurposeCertified Quality
Plate SizeANSI/ SLAS Standard
Unit SizeCase of 120