Thermo Scientific™

Pierce™ Heptafluorobutyric Acid (HFBA), Sequencing Grade

Catalog number: 25003
Have Questions?
Thermo Scientific™

Pierce™ Heptafluorobutyric Acid (HFBA), Sequencing Grade

Catalog number: 25003
Have Questions?
Catalog Number
25003
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
25003100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Heptafluorobutyric Acid (HFBA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent.

Features of HFBA:

Greater than 99.5% purity—allows sensitive, nondestructive peptide detection at low UV wavelengths in peptide separation systems
High-performance packaging—HFBA packaged under nitrogen
Quality control—tested for overall purity, chain-length purity, water content, sulfate content and absorptivity at three different UV-wavelengths

Applications:
• Protein/peptide sequencing
• Protein/peptide solubilizing agent
• Solid-phase peptide synthesis
• Amino acid analysis

Pierce HFBA is manufactured to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment.

Related Products
Pierce™ Heptafluorobutyric Acid (HFBA), HPLC Grade
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
Product Line
Pierce™
Product Size
100 mL
Reagent Type
Ion Pairing Reagent

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text