Thermo Scientific Chemicals

1-Iodododecane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 250090250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

1-Iodododecane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 250090250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 250090250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4292-19-7
IUPAC Name
1-iodododecane
Molecular Formula
C12H25I
InChI Key
GCDPERPXPREHJF-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCI
Appearance (Color)
Clear colorless to light yellow
GC
>=97.5 %
Stabilizer
Copper
Refractive index
1.4825 to 1.4865 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text