1x1 image pixel for data collection
Arecoline hydrobromide, 98+%
Arecoline hydrobromide, 98+%
Arecoline hydrobromide, 98+%
Arecoline hydrobromide, 98+%
Thermo Scientific Chemicals

Arecoline hydrobromide, 98+%

CAS: 300-08-3 | C8H14BrNO2 | 236.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 250130050
Price (USD)
49.65
Special offer
Online exclusive
Ends: 26-Sep-2025
61.10
Save 11.45 (19%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 26-Sep-2025
61.10
Save 11.45 (19%)
Each
Add to cart
Chemical Identifiers
CAS300-08-3
IUPAC Namehydrogen methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate bromide
Molecular FormulaC8H14BrNO2
InChI KeyAXOJRQLKMVSHHZ-UHFFFAOYSA-N
SMILES[H+].[Br-].COC(=O)C1=CCCN(C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Argentometric>=98.0 %
Appearance (Color)White to light yellow
Melting point168°C to 174°C
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0450905Certificate of AnalysisMay 02, 2023250130050, 250131000, 250130250
A0374979Certificate of AnalysisDec 13, 2022250130050, 250131000, 250130250
A0407407Certificate of AnalysisOct 18, 2021250130050, 250131000, 250130250
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection