Thermo Scientific Chemicals

Citric acid, disodium salt sesquihydrate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 250240025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Citric acid, disodium salt sesquihydrate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 250240025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 250240025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6132-05-4
IUPAC Name
tetrasodium bis(2-(carboxymethyl)-2-hydroxybutanedioate) trihydrate
Molecular Formula
C12H18Na4O17
InChI Key
HGPVLOQNBSHYEI-UHFFFAOYNA-J
SMILES
O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CC(O)(CC([O-])=O)C([O-])=O.OC(=O)CC(O)(CC([O-])=O)C([O-])=O
Appearance (Color)
White
Insoluble matter
=<0.01 % (in water)
Appearance (Form)
Crystalline powder or crystals
Titration with HClO4
98.5 to 104.0 %
pH
4.9 to 5.2 (3 % solution)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text