Thermo Scientific Chemicals

Isonicotinic anhydride, 90%, remainder picolinic and nicotinic acid-anhydride, Thermo Scientific Chemicals

Catalog number: 250280050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Isonicotinic anhydride, 90%, remainder picolinic and nicotinic acid-anhydride, Thermo Scientific Chemicals

Catalog number: 250280050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 250280050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7082-71-5
IUPAC Name
pyridine-4-carbonyl pyridine-4-carboxylate
Molecular Formula
C12H8N2O3
InChI Key
ORBIGZZHMAJXAZ-UHFFFAOYSA-N
SMILES
O=C(OC(=O)C1=CC=NC=C1)C1=CC=NC=C1
Appearance (Form)
Crystalline powder
Titration with HClO4
>=88 %
Appearance (Color)
Off-white to beige to light brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text