Thermo Scientific Chemicals

Theophylline, 99+%, anhydrous, Thermo Scientific Chemicals

Catalog number: 250310010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Theophylline, 99+%, anhydrous, Thermo Scientific Chemicals

Catalog number: 250310010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 250310010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-55-9
IUPAC Name
1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C7H8N4O2
InChI Key
ZFXYFBGIUFBOJW-UHFFFAOYSA-N
SMILES
CN1C2=C(NC=N2)C(=O)N(C)C1=O
Infrared spectrum
Conforms
Titration with NaOH
>=99.0 %
Appearance (Color)
White
Melting point
270°C to 274°C
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text