Theophylline, 99+%, anhydrous
Theophylline, 99+%, anhydrous
Theophylline, 99+%, anhydrous
Theophylline, 99+%, anhydrous
Thermo Scientific Chemicals

Theophylline, 99+%, anhydrous

CAS: 58-55-9 | C7H8N4O2 | 180.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 250310050
Price (USD)
30.65
Special offer
Online exclusive
35.50
Save 4.85 (14%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
35.50
Save 4.85 (14%)
Each
Add to cart
Chemical Identifiers
CAS58-55-9
IUPAC Name1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC7H8N4O2
InChI KeyZFXYFBGIUFBOJW-UHFFFAOYSA-N
SMILESCN1C2=C(NC=N2)C(=O)N(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=99.0 %
Appearance (Color)White
Melting point270°C to 274°C
Appearance (Form)Crystalline powder
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link