Thermo Scientific Chemicals

Triisopropylsilyl-trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: 250700100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Triisopropylsilyl-trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: 250700100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 250700100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80522-42-5
IUPAC Name
tris(propan-2-yl)silyl trifluoromethanesulfonate
Molecular Formula
C10H21F3O3SSi
InChI Key
LHJCZOXMCGQVDQ-UHFFFAOYSA-N
SMILES
CC(C)[Si](OS(=O)(=O)C(F)(F)F)(C(C)C)C(C)C
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow to light brown
Titration with NaOH
>=96.0 %
Appearance (Form)
Liquid
Refractive index
1.4130 to 1.4160 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text