Thermo Scientific Chemicals

alpha-Cyclopentylmandelic acid, 98%, Thermo Scientific Chemicals

Catalog number: 250970050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

alpha-Cyclopentylmandelic acid, 98%, Thermo Scientific Chemicals

Catalog number: 250970050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 250970050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
427-49-6
IUPAC Name
2-cyclopentyl-2-hydroxy-2-phenylacetic acid
Molecular Formula
C13H16O3
InChI Key
WFLUEQCOAQCQLP-UHFFFAOYNA-N
SMILES
OC(=O)C(O)(C1CCCC1)C1=CC=CC=C1
Appearance (Color)
White to beige to pink
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
144°C to 150°C
Titration with NaOH
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text