Search Thermo Fisher Scientific
Boost productivity and data quality during volatiles analysis with the highly reliable, robust, and compact TriPlus 500 Headspace Autosampler.
Catalog Number | Description | Software | Injection Mode |
---|---|---|---|
25118200 | Vial tray for upgrade from 120 to 240 vial capacity | Thermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software | Vial Tray Only |
MI-25118000 | Vial Loader and Vial Tray for Upgrade from 12 to 120 Vial Capacity | Thermo Scientific™ Chromeleon CDS, Thermo Scientific™ TraceFinder, and Thermo Scientific™ XCalibur Software | Vial Tray Only |
MI-25118100 | TriPlus™ 500 Headspace Autosampler with Direct Column Connection | Thermo Scientific™ Chromeleon CDS, Thermo Scientific™ TraceFinder, and Thermo Scientific™ XCalibur Software | Standard; Multiple Headspace Extraction (MHE); Multiple Headspace Injection (MHI) |
MI-25118300 | TriPlus™ 500 Headspace Autosampler with Transfer Line | Thermo Scientific™ Chromeleon CDS, Thermo Scientific™ TraceFinder, and Thermo Scientific™ XCalibur Software | Standard; Multiple Headspace Extraction (MHE); Multiple Headspace Injection (MHI) |
Powered by a completely new design, the Thermo Scientific™ TriPlus™ 500 Headspace Autosampler delivers reliability and robustness for testing laboratories performing volatiles analysis. Valve-and-loop technology and other innovative features ensure that this autosampler streamlines daily workflows, saving time with reliable unattended operation, optimizing sample throughput with expanded vial capability, and facilitating method portability with powerful software. Smartly integrated with the Thermo Scientific™ TRACE™ 1600 Series GC, this modular platform provides today's analytical testing and services laboratories with the productivity needed to succeed.
Reliability for analytical testing and services laboratories and versatility for research and method development.
The new design of the TriPlus 500 Headspace Autosampler incorporates innovative features for highly accurate analytical results, including:
New pneumatic controls for high precision and robustness
Best sample integrity and ease-of-use via direct column connection
Compact yet scalable design for better laboratory efficiency and productivity
Comprehensive HS-GC configuration solution
Software and system checks for compliance and data integrity
Discover how Dr. Shelton Gong, a chemist at the Extractable and Leachable testing laboratory at SGS, uses the TriPlus 500 Headspace autosampler and HRAM Orbitrap GC-MS to optimize his E&L workflow, assessing and quantifying potentially dangerous untargeted volatile extractables in polymeric materials down to trace levels.
Share catalog number, name or link