Thermo Scientific Chemicals

Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 252531000
100 mL, Each
Thermo Scientific Chemicals

Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 252531000
100 mL, Each
Quantity
Catalog number: 252531000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14593-46-5, 96-47-9
Appearance
May develop some turbidity or precipitate
Appearance (Form)
Solution
Titration with HClO4
>=30 %
Appearance (Color)
Yellow to orange to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text