Sodium tert-butoxide, 99%, pure, Thermo Scientific Chemicals
Sodium tert-butoxide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium tert-butoxide, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
100 g
500 g
Packaging:
View all
Plastic Drum
Catalog number 252550025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
IUPAC Namesodium 2-methylpropan-2-olate
Molecular FormulaC4H9NaO
InChI KeyMFRIHAYPQRLWNB-UHFFFAOYSA-N
SMILES[Na+].CC(C)(C)[O-]
Molecular Weight (g/mol)96.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
NaOH + Na2CO3 (as NaOH)=<1.5 %
Titration with HCl98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only