Trypsin-EDTA (0.05%), phenol red
Save up to 29% on your order of cell culture products. View promotion details >
Trypsin-EDTA (0.05%), phenol red
Gibco™

Trypsin-EDTA (0.05%), phenol red

Gibco Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestiveRead more
Promo ImagePromotion
Promo code:RCELL25
Save up to 29% across the cell culture workflow - research begins with cells
From now until Dec 31, get up to 29% off media, sera, buffers, reagents and other cell culture products to advance your researchLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
25300054Promo Image100 mL
25300062Promo Image500 mL
25300120Promo Image20 x 100 mL
Catalog number 25300054
Price (USD)
18.65
Online Exclusive
20.64
Save 1.99 (10%)
Each
In stock
Add to cart
Quantity:
100 mL
Customize this product
Price (USD)
18.65
Online Exclusive
20.64
Save 1.99 (10%)
Each
Add to cart
Gibco Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements. See the complete range of Gibco trypsin solutions and recommended dissociation conditions.

We offer a variety of trypsin formulations and animal origin-free TrypLE reagent that feature:

Quality Testing
Documented Traceability
Dual-site cGMP Manufacturing

This trypsin solution is modified as follows:

With
• EDTA
• Phenol Red

The complete formulation is available.

Quality Testing
Gibco Trypsin solutions are tested for pH, osmolality, sterility, and performance. In addition, the raw materials are verified for e-beam irradiation and precursor lots are tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis.

Documented Traceability
We can provide detailed documentation to meet your regulatory needs. Gibco Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.

cGMP Manufacturing and Quality System
For supply chain continuity, we manufacture Gibco Trypsin-EDTA (0.05%), phenol red at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.
Specifications
Cell TypeMammalian Cells
ChelatorsEDTA
Concentration1 X
Osmolality270 - 310 mOsm/kg
Product LineGibco™
Quantity100 mL
Shelf Life24 Months
Shipping ConditionWet ice
SourceAnimal Origin
Tested ForIn vitro Bioassay
ClassificationAnimal Origin
FormLiquid
Product TypeCell Culture Dissociation Reagent
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesPhenol Red, EDTA
pH7.1 to 8
Unit SizeEach
Contents & Storage
Storage conditions: -5°C to -20°C
Shelf life: 24 months from date of manufacture
four-plex-image-COS7-live-cell promo pod

Follow the 5 steps to publication-quality fixed-cell imaging.
Download your free 5-step guide today ›

Frequently asked questions (FAQs)

How do you isolate cells from tissue?

Dissociation of cells from Primary Tissue

TRYPSIN
1. After dissecting off unusable tissue, mince the remaining tissue into 3 to 4 mm pieces with a sterile scalpel or scissors. Wash the tissue pieces by resuspending in a balanced salt solution without calcium and magnesium. Allow the tissue pieces to settle, and remove the supernatant. Repeat the wash 2 or 3 times.
2. Place the container with the tissue pieces on ice, and remove any remaining supernatant. Add 0.25% trypsin in a balanced salt solution without calcium or magnesium (1 ml of trypsin for every 100 mg of tissue).
3. Incubate at 4°C for 6 to 18 h to maximize penetration of the enzyme with little trypsin activity.
4. Decant and discard the trypsin from the tissue pieces. Incubate the tissue pieces with residual trypsin at 37°C for 20 to 30 min.
5. Add warm, complete media to the tissue pieces and gently disperse the tissue by pipetting. If using a serum-free medium, also add soybean trypsin inhibitor.
6. Filter the cell suspension through sterile, stainless steel mesh (100 to 200 µm) to completely disperse any remaining tissue. Count and seed the cells for culture.

COLLAGENASE
1. Mince tissue into 3 to 4 mm pieces with a sterile scalpel or scissors. Wash the tissue pieces several times with Hanks' Balanced Salt Solution (HBSS).
2. Add collagenase (50 to 200 U/ml in HBSS).
3. Incubate at 37°C for 4 to 18 h. Addition of 3 mM CaCl2 increases the efficiency of dissociation.
4. Filter the cell suspension through a sterile stainless steel or nylon mesh to separate the dispersed cells and tissue fragments from the larger pieces. Fresh collagenase can be added to the fragments if further disaggregation is required.
5. Wash suspension several times by centrifugation in HBSS.
6. Resuspend the pellet in culture medium. Count and seed the cells for culture.

DISPASE
1. Mince tissue into 3 to 4 mm pieces with a sterile scalpel or scissors. Wash the tissue pieces several times in a calcium and magnesium-free balanced salt solution.
2. Add dispase (0.6 to 2.4 U/ml in calcium and magnesium-free balanced salt solution).
3. Incubate at 37°C for 20 min to several hours.
4. Filter the cell suspension through a sterile, stainless steel or nylon mesh to separate the dispersed cells and tissue fragments from the larger pieces. Fresh dispase can be added to the fragments if further disaggregation is required.
5. Wash suspension several times by centrifugation in the balanced salt solution.
6. Resuspend the pellet in culture medium. Count and seed the cells for culture.

REFERENCE:
Freshney, R. (1987) Culture of Animal Cells: A Manual of Basic Technique, p. 117, Alan R. Liss, Inc., New York.

Does Diploid Growth Serum-Reduced Medium (SRM) contain L-glutamine?

The basal Diploid Growth Serum-Reduced Medium already contains 6 mM L-glutamine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do I have to adjust my infection parameters when using Diploid Production Serum-Free Medium (SFM)?

We recommend evaluating performance with Diploid Production Serum-Free Medium using current conditions, however, multiplicity of infection, time of infection, and time of harvest may be different than with conventional media. Closely monitor cells for cytopathic effect and evaluate viral titers at multiple time points.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can Diploid Growth Serum-Reduced Medium (SRM) be supplemented with different types of serum?

Diploid Growth Serum-Reduced Medium can be supplemented with Fetal Bovine Serum, Newborn Calf Serum, Donor Bovine Serum with Iron, or Bovine Serum.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which cell lines can I culture in Diploid Growth Serum-Reduced Medium (SRM)?

Diploid Growth Serum-Reduced Medium has been evaluated with fibroblast cell lines, including MRC-5, WI-38, IMR-90, BS-2, and CEF cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link