Thermo Scientific Chemicals

Promethazine hydrochloride

Catalog number: 254580050
5 g, Each
Thermo Scientific Chemicals

Promethazine hydrochloride

Catalog number: 254580050
5 g, Each
Quantity
Catalog number: 254580050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
58-33-3
IUPAC Name
hydrogen dimethyl[1-(10H-phenothiazin-10-yl)propan-2-yl]amine chloride
Molecular Formula
C17H21ClN2S
InChI Key
XXPDBLUZJRXNNZ-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CC(CN1C2=CC=CC=C2SC2=CC=CC=C12)N(C)C
Appearance (Form)
Powder
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Loss on drying
=<0.5 % (105°C, 4 hr)
Solubility
(10% in water) Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text