Promethazine hydrochloride
Promethazine hydrochloride
Promethazine hydrochloride
Promethazine hydrochloride
Thermo Scientific Chemicals

Promethazine hydrochloride

CAS: 58-33-3 | C17H20N2S·HCl | 320.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 254581000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS58-33-3
IUPAC Namehydrogen dimethyl[1-(10H-phenothiazin-10-yl)propan-2-yl]amine chloride
Molecular FormulaC17H21ClN2S
InChI KeyXXPDBLUZJRXNNZ-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(CN1C2=CC=CC=C2SC2=CC=CC=C12)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Powder
Loss on drying=<0.5 % (105°C, 4 hr)
Solubility(10% in water) Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link