Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 255210025
Price (USD)/ Each
88.65
Online exclusive
98.30 
Save 9.65 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
98.30 
Save 9.65 (10%)
Add to cart
Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals
Catalog number255210025
Price (USD)/ Each
88.65
Online exclusive
98.30 
Save 9.65 (10%)
-
Add to cart
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Chloride (Cl)=<50 ppm
Appearance (Color)Clear colorless
Titration with HCl>=25 % (As NH3)
Sulfate (SO4)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only