Thermo Scientific Chemicals

L(+)-Gulonic acid γ-lactone, 95+%, Thermo Scientific Chemicals

Catalog number: 255571000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

L(+)-Gulonic acid γ-lactone, 95+%, Thermo Scientific Chemicals

Catalog number: 255571000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 255571000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1128-23-0
IUPAC Name
(3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
Molecular Formula
C6H10O6
InChI Key
SXZYCXMUPBBULW-SKNVOMKLSA-N
SMILES
OC[C@H](O)[C@H]1OC(=O)[C@@H](O)[C@H]1O
Appearance (Form)
Crystalline powder or crystals and/or chunks
HPLC
>=95.0 %
Specific optical rotation
+51° to +58° (20°C, 589 nm) (c=4, water)
Appearance (Color)
White to light beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text