3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals

3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 255795000
Price (USD)/ Each
303.65
Online exclusive
357.00 
Save 53.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
303.65
Online exclusive
357.00 
Save 53.35 (15%)
Add to cart
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
Catalog number255795000
Price (USD)/ Each
303.65
Online exclusive
357.00 
Save 53.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name3-chlorobenzene-1-carboperoxoic acid
Molecular FormulaC7H5ClO3
InChI KeyNHQDETIJWKXCTC-UHFFFAOYSA-N
SMILESOOC(=O)C1=CC=CC(Cl)=C1
Molecular Weight (g/mol)172.56
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
3-Chlorobenzoic acid=<12 %
HPLCdi-m-Chlorobenzoyl peroxide: < 0.1% Typical
HPLC=<0.2 % di-m-Chlorobenzoyl peroxide
Titration Iodometric70 to 75 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only