1,10-Dichlorodecane, 98%, Thermo Scientific Chemicals
1,10-Dichlorodecane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,10-Dichlorodecane, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 255930250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2162-98-3
IUPAC Name1,10-dichlorodecane
Molecular FormulaC10H20Cl2
InChI KeyRBBNTRDPSVZESY-UHFFFAOYSA-N
SMILESClCCCCCCCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only