Bacto Brain Heart Infusion, porcine, is used for cultivating a wide variety of microorganisms. It was developed as an alternative to the classical Brain Heart Infusion formula and replaces calf brain and beef heart with pork brain and heart. Bacto Brain Heart Infusion, porcine, was formulated with no bovine components to minimize bovine spongiform encephalopathy (BSE) risk.
Specifications
Product LineBacto™
Quantity10 kg
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.