3,3-Dimethylbutyraldehyde, Thermo Scientific Chemicals
3,3-Dimethylbutyraldehyde, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Dimethylbutyraldehyde, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 257260250
Price (USD)/ Each
868.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
868.00
Add to cart
3,3-Dimethylbutyraldehyde, Thermo Scientific Chemicals
Catalog number257260250
Price (USD)/ Each
868.00
-
Add to cart
Chemical Identifiers
IUPAC Name3,3-dimethylbutanal
Molecular FormulaC6H12O
InChI KeyLTNUSYNQZJZUSY-UHFFFAOYSA-N
SMILESCC(C)(C)CC=O
Molecular Weight (g/mol)100.16
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3960 to 1.3980 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only