Thermo Scientific Chemicals

2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution, Thermo Scientific Chemicals

Catalog number: 257740025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution, Thermo Scientific Chemicals

Catalog number: 257740025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 257740025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3811-73-2, 7732-18-5
Titration with HClO4
40 to 42 %
Appearance (Color)
Yellow to amber
pH
8.5 to 10.5 (10 % at 20°C)
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text