Thermo Scientific Chemicals

Isobutyric anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 257785000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Isobutyric anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 257785000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 257785000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
97-72-3
IUPAC Name
2-methylpropanoyl 2-methylpropanoate
Molecular Formula
C8H14O3
InChI Key
LSACYLWPPQLVSM-UHFFFAOYSA-N
SMILES
CC(C)C(=O)OC(=O)C(C)C
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4050 to 1.4070 (20°C, 589 nm)
GC
=<1.0 % (Isobutyric acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text