Methyl cellulose, viscosity 4000 cP (2% solution in water)
Methyl cellulose, viscosity 4000 cP (2% solution in water)
Methyl cellulose, viscosity 4000 cP (2% solution in water)
Methyl cellulose, viscosity 4000 cP (2% solution in water)
Thermo Scientific Chemicals

Methyl cellulose, viscosity 4000 cP (2% solution in water)

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 258111000
Price (USD)
39.65
Online Exclusive
43.80
Save 4.15 (9%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.80
Save 4.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS9004-67-5
IUPAC Name3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular FormulaC20H38O11
InChI KeyYLGXILFCIXHCMC-UHFFFAOYNA-N
SMILESCOCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellowish-white to greyish-white
Infrared spectrumConforms
Viscosity3000 to 5600 cP (2 % in water at 20°C)
Appearance (Form)Powder
Loss on drying=<5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473568Certificate of AnalysisJun 26, 2025258115000, 258110025, 258111000
A0455040Certificate of AnalysisAug 25, 2023258115000, 258110025, 258111000
A0398630Certificate of AnalysisDec 15, 2022258115000, 258110025, 258111000
A0409005Certificate of AnalysisDec 13, 2022258115000, 258110025, 258111000
A0413771Certificate of AnalysisDec 13, 2022258115000, 258110025, 258111000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection