Thermo Scientific Chemicals

Tetracosanol, 99+%, Thermo Scientific Chemicals

Catalog number: 258151000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

Tetracosanol, 99+%, Thermo Scientific Chemicals

Catalog number: 258151000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 258151000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-51-4
IUPAC Name
tetracosan-1-ol
Molecular Formula
C24H50O
InChI Key
TYWMIZZBOVGFOV-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCCCCO
Appearance (Color)
White
Infrared spectrum
Conforms
GC
>=99.0 %
Appearance (Form)
Glistening powder or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text