Tricosanol, 99+%, Thermo Scientific Chemicals
Tricosanol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tricosanol, 99+%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number 258161000
Price (USD)/ Each
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3133-01-5
IUPAC Nametricosan-1-ol
Molecular FormulaC23H48O
InChI KeyFPLNRAYTBIFSFW-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Glistening crystalline powder or flakes
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only