Thermo Scientific Chemicals

Hexacosanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 258210010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Hexacosanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 258210010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 258210010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-46-7
IUPAC Name
hexacosanoic acid
Molecular Formula
C26H52O2
InChI Key
XMHIUKTWLZUKEX-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Infrared spectrum
Conforms
Appearance (Form)
Glistening powder
Appearance (Color)
White
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text