N-2-Acetylguanine, 99%, Thermo Scientific Chemicals
N-2-Acetylguanine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-2-Acetylguanine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 258400010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19962-37-9
IUPAC NameN-(6-oxo-6,7-dihydro-3H-purin-2-yl)acetamide
Molecular FormulaC7H7N5O2
InChI KeyMXSMRDDXWJSGMC-UHFFFAOYSA-N
SMILESCC(=O)NC1=NC(=O)C2=C(N1)N=CN2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Titration with HClO4>=98.5 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only