Acetonitrile, 99.5%, for analysis, Thermo Scientific Chemicals
Acetonitrile, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile, 99.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 258560010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.4 %
Water=<0.1 % (Coulometric)
Lead (Pb)=<2 ppm
Iron (Fe)=<2 ppm
Copper (Cu)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only