1x1 image pixel for data collection
Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Thermo Scientific Chemicals

Buffer solution pH 4, Citrate buffer

CAS: 68-04-2 | C8H5KO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
5 L
Catalog number 258585000
Price (USD)
47.65
Special offer
Online exclusive
Ends: 26-Sep-2025
59.20
Save 11.55 (20%)
Each
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 26-Sep-2025
59.20
Save 11.55 (20%)
Each
Add to cart
Chemical Identifiers
CAS68-04-2
SpecificationsSpecification SheetSpecification Sheet
pH3.98 to 4.02 (at 20°C) composition: citrate-NaCl-NaOH
Appearance (Color)Clear colorless
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH buffer
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link