Thermo Scientific Chemicals

Buffer solution pH 7, Phosphate buffer, Thermo Scientific Chemicals

Catalog number: 258590010
1 L, Plastic bottle, Each
Thermo Scientific Chemicals

Buffer solution pH 7, Phosphate buffer, Thermo Scientific Chemicals

Catalog number: 258590010
1 L, Plastic bottle, Each
Quantity
Packaging
Catalog number: 258590010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7558-79-4, 7732-18-5
Appearance (Form)
Solution
Appearance (Color)
Clear colorless
pH
6.98 to 7.02 (at 20°C) composition: 3.5 g
pH
potassium dihydrogen phosphate and 12.5 g
pH
di-sodium hydrogen phosphate per liter

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text