Buffer solution pH 10, Carbonate buffer
Buffer solution pH 10, Carbonate buffer
Thermo Scientific Chemicals

Buffer solution pH 10, Carbonate buffer

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Catalog number 258600025
Price (USD)
113.65
Special offer
Online exclusive
Ends: 26-Sep-2025
140.00
Save 26.35 (19%)
Each
-
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 26-Sep-2025
140.00
Save 26.35 (19%)
Each
Add to cart
Specifications
CAS1332-57-6
CAS144-55-8
CAS7732-18-5
Chemical Name or MaterialBuffer solution pH 10
ColorColorless
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH buffer
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470613Certificate of AnalysisMay 17, 2025258605000, 258600010, 258600025
A0460004Certificate of AnalysisFeb 06, 2025258605000, 258600010, 258600025
A0460004Certificate of AnalysisFeb 08, 2024258605000, 258600010, 258600025
B00U0533Certificate of AnalysisOct 20, 2023258605000, 258600010, 258600025
A0456040Certificate of AnalysisSep 13, 2023258605000, 258600010, 258600025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection